missing translation for 'onlineSavingsMsg'
Learn More

Cantrodifene, TRC

Product Code. 30407052
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407052 50 mg 50mg
30404904 250 mg -
30405136 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407052 Supplier Toronto Research Chemicals Supplier No. TRCI90569050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19881-18-6
Molecular Formula C13H8N2O3S
Molecular Weight (g/mol) 272.27
Synonym Nitroscanate, 1-Isothiocyanato-4-(4-nitrophenoxy)benzene (ACI), Isothiocyanic acid, p-(p-nitrophenoxy)phenyl ester (8CI), 4-(4-Nitrophenoxy)phenyl isothiocyanate, 4-Nitro-4'-isothiocyanatodiphenyl ether, Cantrodifene, Lopatol, Nitroscanate
IUPAC Name 1-isothiocyanato-4-(4-nitrophenoxy)benzene
SMILES C1=C(C=CC(=C1)OC2=CC=C(C=C2)[N+](=O)[O-])N=C=S

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H8N2O3S/c16-15(17)11-3-7-13(8-4-11)18-12-5-1-10(2-6-12)14-9-19/h1-8H
Formula Weight 272.0256
Chemical Name or Material 4-Isothiocyanato-4'-nitrodiphenyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.