CAS RN 19881-18-6
CAS RN 19881-18-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cantrodifene, TRC
CAS: 19881-18-6 Molecular Formula: C13H8N2O3S Molecular Weight (g/mol): 272.27 Synonym: Nitroscanate, 1-Isothiocyanato-4-(4-nitrophenoxy)benzene (ACI),Isothiocyanic acid, p-(p-nitrophenoxy)phenyl ester (8CI),4-(4-Nitrophenoxy)phenyl isothiocyanate,4-Nitro-4'-isothiocyanatodiphenyl ether,Cantrodifene,Lopatol,Nitroscanate IUPAC Name: 1-isothiocyanato-4-(4-nitrophenoxy)benzene SMILES: C1=C(C=CC(=C1)OC2=CC=C(C=C2)[N+](=O)[O-])N=C=S