missing translation for 'onlineSavingsMsg'
Learn More

Calconcarboxylic Acid, Supelco™

Product Code. 15814450
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814450 25 g 25g
15804450 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15814450 Supplier Supelco Supplier No. 1045950025
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Metal indicator

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3737-95-9

Specifications

Melting Point 300°C
MDL Number MFCD00004078
Synonym Calconcarboxylic acid, Patton and Reeders Reagent, 2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-naphthalene-3-carboxylic acid, Calconcarboxylic acid
Quantity 25 g
Molecular Weight (g/mol) 474.44
Grade Research
Chemical Name or Material Calconcarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.