accessibility menu, dialog, popup

UserName

OHOHNNOSOHOOOH(Z)

CAS RN 3737-95-9

IUPAC Name: 3-hydroxy-4-{2-[(1Z)-2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-2-carboxylic acid
Synonyms: 3-hydroxy-4-{2-[(1Z)-2-oxo-4-sulfonaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-2-carboxylic acid
Molecular Weight (g/mol): 438.41
Molecular Formula: C21H14N2O7S
InChi Key: ULIVOAKVRBXKKS-PYCFMQQDSA-N
SMILES: OC(=O)C1=C(O)C(N\N=C2/C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (3)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)