Learn More
Calcium D-saccharate tetrahydrate, 98%
Product Code. 15448519
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448519 100 g 100g
15438519 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15448519 Supplier Thermo Scientific Alfa Aesar Supplier No. J66947.22
Request Bulk or Custom Format
CAS: 5793-89-5 | C6H16CaO12 | 320.26 g/mol
D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.
Solubility
Soluble in water(0.43g/L).
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5793-89-5
Molecular Formula C6H16CaO12
Molecular Weight (g/mol) 320.26
MDL Number MFCD00150704
InChI Key ZQWFSIZRQANUDA-WQMSYZFBSA-L
Synonym calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrate
PubChem CID 16211545
IUPAC Name calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate
SMILES C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2]
Specifications
Color White
Physical Form Powder
Quantity 100 g
Beilstein 3821334
Merck Index 14,1700
Solubility Information Soluble in water(0.43g/L).
Formula Weight 320.26
Percent Purity 98%
Odor Odorless
Chemical Name or Material Calcium D-saccharate tetrahydrate
compliance-icons
Product Identifier
  • Calcium D-saccharate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Calcium D-saccharatetetrahydrate