accessibility menu, dialog, popup

UserName

CAS RN 5793-89-5

H2OH2OH2OH2OCaOH(S)(R)OH(R)OHOO(S)OHOO

CAS RN 5793-89-5

IUPAC Name: calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
Synonyms: calcium allarate tetrahydrate
Molecular Weight (g/mol): 320.26
Molecular Formula: C6H16CaO12
InChi Key: ZQWFSIZRQANUDA-FARGFPGJSA-L
SMILES: O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results