missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C4.4A/LYPD3 Monoclonal specifically detects C4.4A/LYPD3 in Human samples. It is validated for ELISA/Flow Cytometry/Immunocytochemistry/Immunofluorescence/Western Blot/REMOVE.
Specifications
Specifications
| Antigen | C4.4A/LYPD3 |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 213 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human C4.4A/LYPD3 (NP_055215.2; Met 1-His 286). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?