missing translation for 'onlineSavingsMsg'
Learn More

Butyrylglycine, TRC

Product Code. 30134721
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134721 10 g 10g
30005211 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134721 Supplier Toronto Research Chemicals Supplier No. TRCB69377010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20208-73-5
Molecular Formula C6 H11 N O3
Molecular Weight (g/mol) 145.16
Synonym Glycine, N-(1-oxobutyl)- (9CI, ACI), Glycine, N-butyryl- (6CI, 8CI), N-(1-Oxobutyl)glycine (ACI), (Butyrylamino)acetic acid, 2-(Butanoylamino)acetic acid, 2-Butanamidoacetic acid, Butyrylglycine, N-Butanoylglycine, N-Butyrylglycine
IUPAC Name 2-(butanoylamino)acetic acid
SMILES CCCC(=O)NCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H11NO3/c1-2-3-5(8)7-4-6(9)10/h2-4H2,1H3,(H,7,8)(H,9,10)
Formula Weight 145.0739
Percent Purity >95%
Chemical Name or Material Butyrylglycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.