CAS RN 20208-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Butyrylglycine, TRC
CAS: 20208-73-5 Molecular Formula: C6 H11 N O3 Molecular Weight (g/mol): 145.16 Synonym: Glycine, N-(1-oxobutyl)- (9CI, ACI),Glycine, N-butyryl- (6CI, 8CI),N-(1-Oxobutyl)glycine (ACI),(Butyrylamino)acetic acid,2-(Butanoylamino)acetic acid,2-Butanamidoacetic acid,Butyrylglycine,N-Butanoylglycine,N-Butyrylglycine IUPAC Name: 2-(butanoylamino)acetic acid SMILES: CCCC(=O)NCC(=O)O