Learn More
Butyl Methanesulfonate, TRC
Product Code. 30010232
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010232 500 mg 500mg
30008959 5 g 5g
30008601 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010232 Supplier Toronto Research Chemicals Supplier No. TRCB694095500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1912-32-9
Molecular Formula C5 H12 O3 S
Molecular Weight (g/mol) 152.21
Synonym Methanesulfonic acid butyl ester, Butyl mesylate, Butyl methanesulfonate, Methanesulfonic acid n-butyl ester, NSC 36060, n-Butyl methanesulfonate
IUPAC Name butyl methanesulfonate
SMILES CCCCOS(=O)(=O)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C5H12O3S/c1-3-4-5-8-9(2,6)7/h3-5H2,1-2H3
Formula Weight 152.0507
Percent Purity >95%
Chemical Name or Material Butyl methanesulfonate
Grade GC