CAS RN 1912-32-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Butyl Methanesulfonate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Butyl Methanesulfonate, TRC
CAS: 1912-32-9 Molecular Formula: C5 H12 O3 S Molecular Weight (g/mol): 152.21 Synonym: Methanesulfonic acid butyl ester,Butyl mesylate,Butyl methanesulfonate,Methanesulfonic acid n-butyl ester,NSC 36060,n-Butyl methanesulfonate IUPAC Name: butyl methanesulfonate SMILES: CCCCOS(=O)(=O)C