Learn More
Bromoethane, Sigma-Aldrich™
Product Code. 15898430
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898430 250 mL 250mL
15808440 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15898430 Supplier Sigma Aldrich Supplier No. 8008710250
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74-96-4
Grade Research
Molecular Weight (g/mol) 108.97
Specifications
Melting Point -118°C
Density 1.46 g/cm3 at 20°C
Boiling Point 37°C to 39°C/1013 hPa
MDL Number MFCD00000232
Quantity 250 mL
Synonym Bromoethane, Ethyl bromide, Bromoethane