CAS RN 74-96-4
CAS RN 74-96-4
IUPAC Name:
bromoethane
Synonyms:
bromoethane
Molecular Weight (g/mol):
108.97
Molecular Formula:
C2H5Br
InChi Key:
RDHPKYGYEGBMSE-UHFFFAOYSA-N
SMILES:
CCBr
1
–
2
of
2
results
1
Bromoethane, 98%, pure
CAS: 74-96-4 Molecular Formula: C2H5Br MDL Number: MFCD00000232 InChI Key: RDHPKYGYEGBMSE-UHFFFAOYSA-N Synonym: ethyl bromide,ethane, bromo,1-bromoethane,monobromoethane,bromic ether,hydrobromic ether,bromure d'ethyle,ethylbromide,etylu bromek,halon 2001 PubChem CID: 6332 IUPAC Name: bromoethane SMILES: CCBr