Learn More
Bromochloroiodomethane, TRC
Product Code. 30138156
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138156 100 mg 100mg
17904035 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138156 Supplier Toronto Research Chemicals Supplier No. TRCB682375100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34970-00-8
Molecular Formula CHBrClI
Molecular Weight (g/mol) 255.28
Synonym Iodobromochloromethane
IUPAC Name bromo-chloro-iodomethane
SMILES ClC([H])(I)Br
Specifications
Quantity 100 mg
InChI Formula InChI=1S/CHBrClI/c2-1(3)4/h1H
Formula Weight 253.7995
Chemical Name or Material Bromochloroiodomethane