CAS RN 34970-00-8
CAS RN 34970-00-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bromochloroiodomethane, TRC
CAS: 34970-00-8 Molecular Formula: CHBrClI Molecular Weight (g/mol): 255.28 Synonym: Iodobromochloromethane IUPAC Name: bromo-chloro-iodomethane SMILES: ClC([H])(I)Br