Learn More
Novus Biologicals™ Brain Tissue Slides (Meningioma Transitional)- Paraffin
Product Code. 18103963 Shop All Bio Techne Products
Change view
Click to view available options
Species:
Human
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species unitSize
18103963 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18103963 Supplier Novus Biologicals™ Supplier No. NBP230254
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Type Brain Tissue Slides (Meningioma Transitional)
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Meningioma Transitional