missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borrelia Burgdorferi Flagellar Antigen Antibody (7G11/6802), Alexa Fluor™ 750, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Borrelia Burgdorferi Flagellar Antigen Monoclonal specifically detects Borrelia Burgdorferi Flagellar Antigen in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Borrelia Burgdorferi Flagellar Antigen |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 7G11/6802 |
| Conjugate | Alexa Fluor 750 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease |
| Host Species | Mouse |
| Immunogen | Native |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?