missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron Trifluoride-diethyl Ether Complex, Sigma-Aldrich™
Spezifikation
Spezifikation
| Melting Point | -58°C |
| Density | 1.13 g/cm3 at 20°C |
| MDL Number | MFCD00013194 |
| Quantity | 1 L |
| Synonym | Boron trifluoride-diethyl ether complex, Boron trifluoride-ethyl etherate, Boron trifluoride-diethyl ether complex |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?