accessibility menu, dialog, popup

UserName

CAS RN 109-63-7

BFFFOH3CH3C

CAS RN 109-63-7

IUPAC Name: ethoxyethane; trifluoroborane
Synonyms: boron trifluoride; diethyl ether
Molecular Weight (g/mol): 141.93
Molecular Formula: C4H10BF3O
InChi Key: KZMGYPLQYOPHEL-UHFFFAOYSA-N
SMILES: FB(F)F.CCOCC
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)