missing translation for 'onlineSavingsMsg'
Learn More

Bomyl, TRC

Product Code. 30134198
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134198 100 mg 100mg
30136077 500 mg 500mg
30012090 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134198 Supplier Toronto Research Chemicals Supplier No. TRCB675250100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-10-1
Molecular Formula C9H15O8P
Molecular Weight (g/mol) 282.18
Synonym 3-[(Dimethoxyphosphinyl)oxy]-2-pentenedioic Acid Dimethyl Ester, Dimethyl 3-hydroxyglutaconate dimethyl phosphate, General Chemicals 3707, Dimethyl-1,3-di(carbomethoxy)-1-propen-2-yl phosphate, ENT 24,833, GC 3707
IUPAC Name dimethyl 3-dimethoxyphosphoryloxypent-2-enedioate
SMILES COC(=O)CC(=CC(=O)OC)OP(=O)(OC)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H15O8P/c1-13-8(10)5-7(6-9(11)14-2)17-18(12,15-3)16-4/h5H,6H2,1-4H3
Formula Weight 282.0505
Chemical Name or Material Bomyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.