CAS RN 122-10-1
CAS RN 122-10-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bomyl, TRC
CAS: 122-10-1 Molecular Formula: C9H15O8P Molecular Weight (g/mol): 282.18 Synonym: 3-[(Dimethoxyphosphinyl)oxy]-2-pentenedioic Acid Dimethyl Ester,Dimethyl 3-hydroxyglutaconate dimethyl phosphate,General Chemicals 3707,Dimethyl-1,3-di(carbomethoxy)-1-propen-2-yl phosphate,ENT 24,833,GC 3707 IUPAC Name: dimethyl 3-dimethoxyphosphoryloxypent-2-enedioate SMILES: COC(=O)CC(=CC(=O)OC)OP(=O)(OC)OC