missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BML 111
GREENER_CHOICE

Product Code. 18735319
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735319 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735319 Supplier Tocris Bioscience™ Supplier No. 5488/10
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

FPR2 (lipoxin A4 receptor) agonist

BML 111 is a FPR2 (lipoxin A4 receptor) agonist. Suppresses tumor-related angiogenesis and reduces tumor growth in hepatoma H22 cell bearing mice. Also protects against hemorrhagic shock-induced acute lung injury in rats. Lipoxin A4 analog.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5S,6R)-Methyl 5,6,7-trihydroxyheptanoate
CAS 78606-80-1
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C8H16O5
Formula Weight 192.11
Physical Form Solid
Color White
Product Type Bioactive Small Molecule

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.