missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A Monosulfate Sodium Salt, TRC

Product Code. 30010121
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010121 100 mg 100mg
30005746 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010121 Supplier Toronto Research Chemicals Supplier No. TRCB519560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 847696-37-1
Molecular Formula C15H15NaO5S
Molecular Weight (g/mol) 330.33
Synonym Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), sodium salt (1:1) (ACI), Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), monosodium salt (9CI), Sodium 4-[1-(4-hydroxyphenyl)-1-methylethyl]phenyl sulfate (1:1) (ACI), Bisphenol A monosulfate sodium salt
IUPAC Name sodium;[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl] sulfate
SMILES OC(C=C1)=CC=C1C(C)(C)C2=CC=C(OS(=O)(O[Na])=O)C=C2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H16O5S.Na/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(10-6-12)20-21(17,18)19;/h3-10,16H,1-2H3,(H,17,18,19);/q;+1/p-1
Formula Weight 330.0538
Percent Purity >95%
Chemical Name or Material Bisphenol A Monosulfate Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.