CAS RN 847696-37-1
CAS RN 847696-37-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bisphenol A Monosulfate Sodium Salt, TRC
CAS: 847696-37-1 Molecular Formula: C15H15NaO5S Molecular Weight (g/mol): 330.33 Synonym: Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), sodium salt (1:1) (ACI),Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), monosodium salt (9CI),Sodium 4-[1-(4-hydroxyphenyl)-1-methylethyl]phenyl sulfate (1:1) (ACI),Bisphenol A monosulfate sodium salt IUPAC Name: sodium;[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl] sulfate SMILES: OC(C=C1)=CC=C1C(C)(C)C2=CC=C(OS(=O)(O[Na])=O)C=C2