Learn More
MedChemExpress Bis-Tos-PEG4
Specifications
Specifications
| Molecular Weight (g/mol) | 502.6 g/mol |
| CAS | 37860-51-8 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 100 mg |
| Synonym | PROTAC Linker 16 |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |