Learn More
Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride, 97%, Thermo Scientific™
Product Code. 11481430
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481430 1 g 1g
11491430 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11481430 Supplier Thermo Scientific Alfa Aesar Supplier No. H26892.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32876-92-9
Molecular Formula C18H26Cl2Zr
Molecular Weight (g/mol) 404.53
MDL Number MFCD01073785
InChI Key MHQMSFSHQBJUHE-UHFFFAOYSA-L
Synonym zirconium 4+ chloride 2-tert-butylcyclopenta-1,3-dien-1-ide 1/2/2
PubChem CID 73882475
IUPAC Name 2-tert-butylcyclopenta-1,3-diene;zirconium(4+);dichloride
SMILES CC(C)(C)C1=[C-]CC=C1.CC(C)(C)C1=[C-]CC=C1.[Cl-].[Cl-].[Zr+4]
Specifications
Melting Point 192°C to 194°C
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 404.53
Percent Purity 97%
Chemical Name or Material Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride