CAS RN 32876-92-9
IUPAC Name:
zirconium(4+) bis(2-tert-butylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms:
zirconium(4+) bis(2-tert-butylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol):
404.53
Molecular Formula:
C18H26Cl2Zr
InChi Key:
MHQMSFSHQBJUHE-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].[Zr+4].CC(C)(C)C1=[C-]CC=C1.CC(C)(C)C1=[C-]CC=C1
1
–
1
of
1
results
1
Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride, 97%, Thermo Scientific™
CAS: 32876-92-9 Molecular Formula: C18H26Cl2Zr Molecular Weight (g/mol): 404.53 MDL Number: MFCD01073785 InChI Key: MHQMSFSHQBJUHE-UHFFFAOYSA-L Synonym: zirconium 4+ chloride 2-tert-butylcyclopenta-1,3-dien-1-ide 1/2/2 PubChem CID: 73882475 IUPAC Name: 2-tert-butylcyclopenta-1,3-diene;zirconium(4+);dichloride SMILES: CC(C)(C)C1=[C-]CC=C1.CC(C)(C)C1=[C-]CC=C1.[Cl-].[Cl-].[Zr+4]