Learn More
Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 94%, Thermo Scientific™
Product Code. 11388036
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388036 1000 g 1000g
11368036 50 g 50g
11378036 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11388036 Supplier Thermo Scientific Alfa Aesar Supplier No. L13323.0B
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77-62-3
Molecular Formula C29H40O2
Molecular Weight (g/mol) 420.64
MDL Number MFCD00151797
InChI Key PHXLONCQBNATSL-UHFFFAOYSA-N
Synonym 2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, ionox wsp, nonox wsp, bisalkofen mtsp, unii-7tir91vdi9, 7tir91vdi9, p-cresol, 2,2'-methylenebis 6-1-methylcyclohexyl, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol
PubChem CID 6486
IUPAC Name 2-[[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl]-4-methyl-6-(1-methylcyclohexyl)phenol
SMILES CC1=CC(=C(O)C(CC2=C(O)C(=CC(C)=C2)C2(C)CCCCC2)=C1)C1(C)CCCCC1
Specifications
Melting Point 130°C to 135°C
Odor Odorless
Quantity 1000 g
Beilstein 2313064
Formula Weight 420.64
Percent Purity 94%
Chemical Name or Material Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane