accessibility menu, dialog, popup

UserName

CH3CH3OHH3COHH3C

CAS RN 77-62-3

IUPAC Name: 2-{[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl}-4-methyl-6-(1-methylcyclohexyl)phenol
Synonyms: 2-{[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl}-4-methyl-6-(1-methylcyclohexyl)phenol
Molecular Weight (g/mol): 420.64
Molecular Formula: C29H40O2
InChi Key: PHXLONCQBNATSL-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O)C(CC2=C(O)C(=CC(C)=C2)C2(C)CCCCC2)=C1)C1(C)CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results