missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-bromobenzyl)amine hydrochloride, 99%

Product Code. 11489227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489227 100 g 100g
11469227 5 g 5g
11479227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21318.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 336615-45-3 | C14H14Br2ClN | 391.531 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336615-45-3
Molecular Formula C14H14Br2ClN
Molecular Weight (g/mol) 391.531
MDL Number MFCD01632133
InChI Key KXMULHDKALNZDJ-UHFFFAOYSA-N
Synonym bis 2-bromobenzyl amine hydrochloride, bis 2-bromophenyl methyl amine hydrochloride, acmc-20aloa, bis 2-bromobenzyl ;amine hydrochloride, 2,2'-dibromodibenzylamine hydrochloride, bis-2-bromobenzyl amine hydrochloride, 1-2-bromophenyl-n-2-bromophenyl methyl methanamine-hydrogen chloride 1/1
PubChem CID 24212083
IUPAC Name 1-(2-bromophenyl)-N-[(2-bromophenyl)methyl]methanamine;hydrochloride
SMILES C1=CC=C(C(=C1)CNCC2=CC=CC=C2Br)Br.Cl

Specifications

Melting Point 165°C to 168°C
Quantity 100 g
Formula Weight 391.53
Percent Purity 99%
Chemical Name or Material Bis(2-bromobenzyl)amine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.