accessibility menu, dialog, popup

UserName

NHBrBrHCl

CAS RN 336615-45-3

IUPAC Name: bis[(2-bromophenyl)methyl]amine hydrochloride
Synonyms: bis[(2-bromophenyl)methyl]amine hydrochloride
Molecular Weight (g/mol): 391.53
Molecular Formula: C14H14Br2ClN
InChi Key: KXMULHDKALNZDJ-UHFFFAOYSA-N
SMILES: Cl.BrC1=CC=CC=C1CNCC1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results