Learn More
Bis(2,5-hydroxymethyl)dioxane(Mixture of Diastereomers), TRC
Product Code. 30136255
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136255 1 g 1g
30010889 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136255 Supplier Toronto Research Chemicals Supplier No. TRCB4453001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14236-12-5
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.16
Synonym Bis(2,5-Hydroxymethyl)Dioxane
IUPAC Name [5-(hydroxymethyl)-1,4-dioxan-2-yl]methanol
SMILES OCC1COC(CO)CO1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H12O4/c7-1-5-3-10-6(2-8)4-9-5/h5-8H,1-4H2
Formula Weight 148.0736
Chemical Name or Material Bis(2,5-hydroxymethyl)dioxane(Mixture of Diastereomers)