CAS RN 14236-12-5
CAS RN 14236-12-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(2,5-hydroxymethyl)dioxane(Mixture of Diastereomers), TRC
CAS: 14236-12-5 Molecular Formula: C6H12O4 Molecular Weight (g/mol): 148.16 Synonym: Bis(2,5-Hydroxymethyl)Dioxane IUPAC Name: [5-(hydroxymethyl)-1,4-dioxan-2-yl]methanol SMILES: OCC1COC(CO)CO1