missing translation for 'onlineSavingsMsg'
Learn More

Birch pollen Bet V1 Antibody (BV16), Alexa Fluor™ 594, R&D Systems™

Product Code. 18228198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18228198 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18228198 Supplier Novus Biologicals Supplier No. NBP281093AF594
  • ONLINE EXCLUSIVE PRICE
    690.22 EUR
    606.90 EUR / 0.1mL
    Save 83.32 EUR
    12% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Birch pollen Bet V1
Applications ELISA, Western Blot
Classification Monoclonal
Clone BV16
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Host Species Rabbit
Immunogen Birch pollen Bet V1-A, a major allergen of the European white birch.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody binds to Birch pollen Bet V1.
Target Species Plant
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.