Learn More
Betahistine Dihydrochloride, TRC
Product Code. 17934265
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934265 5 g 5g
17972555 1 g 1g
17963835 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17934265 Supplier Toronto Research Chemicals Supplier No. TRCB3240015G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5579-84-0
Molecular Formula C8 H12 N2 . 2 Cl H
Molecular Weight (g/mol) 209.12
Synonym Betahistine Dihydrochloride, N-Methyl-2-pyridineethanamine hydrochloride (1:2), N-Methyl-2-pyridineethanamine dihydrochloride, 2-[2-(Methylamino)ethyl]pyridine dihydrochloride, Betahistine dihydrochloride, Betahistine hydrochloride, Betaserc, PT 9, Serc, Sinmenier, Vasomotal
IUPAC Name N-methyl-2-pyridin-2-ylethanamine;dihydrochloride
SMILES Cl.Cl.CNCCc1ccccn1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H12N2.2ClH/c1-9-7-5-8-4-2-3-6-10-8;;/h2-4,6,9H,5,7H2,1H3;2*1H
Formula Weight 208.0534
Percent Purity >95%
Chemical Name or Material Betahistine dihydrochloride
Grade HPLC