CAS RN 5579-84-0
CAS RN 5579-84-0
IUPAC Name:
dihydrogen methyl[2-(pyridin-2-yl)ethyl]amine dichloride
Synonyms:
dihydrogen betahistine dichloride
Molecular Weight (g/mol):
209.11
Molecular Formula:
C8H14Cl2N2
InChi Key:
XVDFMHARQUBJRE-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CNCCC1=CC=CC=N1
1
–
2
of
2
results
1
Betahistine Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Betahistine Dihydrochloride, TRC
CAS: 5579-84-0 Molecular Formula: C8 H12 N2 . 2 Cl H Molecular Weight (g/mol): 209.12 Synonym: Betahistine Dihydrochloride,N-Methyl-2-pyridineethanamine hydrochloride (1:2),N-Methyl-2-pyridineethanamine dihydrochloride,2-[2-(Methylamino)ethyl]pyridine dihydrochloride,Betahistine dihydrochloride,Betahistine hydrochloride,Betaserc,PT 9,Serc,Sinmenier,Vasomotal IUPAC Name: N-methyl-2-pyridin-2-ylethanamine;dihydrochloride SMILES: Cl.Cl.CNCCc1ccccn1