Learn More
Benzoic Acid, Supelco™
Product Code. 15883830
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15883830 1 kg 1kg
15863830 100 g 100g
15873830 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15883830 Supplier Supelco Supplier No. 1001361000
 Restricted Item
Estimated Shipment: 28-01-2027
For analysis EMSURE™ ACS, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65-85-0
Molecular Weight (g/mol) 122.12
Specifications
Melting Point 121°C to 123°C
Boiling Point 249°C/1013 hPa
Density 1.321 g/cm3 at 20°C
Quantity 1 kg
MDL Number MFCD00002398
Synonym Benzoic acid, Phenylformic acid, Benzene carboxylic acid, Benzoic acid
Chemical Name or Material Benzoic Acid