CAS RN 65-85-0
Benzoic acid, 99.6%, ACS reagent
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, 99%, extra pure
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, 99.5%, for analysis
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, ACS, 99.5% min
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid, Certified AR for Analysis, Fisher Chemical™
CAS: 65-85-0 Molecular Formula: C7H6O2 MDL Number: 2398
Benzoic acid, 99%
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
HYDRANAL™ - Benzoic acid, Solstice
Buffer substance for Karl Fischer titration
Benzoic Acid, Supelco™
CAS: 65-85-0 Molecular Weight (g/mol): 122.12
Benzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Benzoic Acid, TRC
CAS: 65-85-0 Molecular Formula: C7 H6 O2 Molecular Weight (g/mol): 122.12 Synonym: Benzenecarboxylic acid,Benzeneformic acid,Benzenemethanoic acid,Carboxybenzene,Dracylic acid,E 210,HA 1,HA 1 (acid),MENNO-Florades,NSC 149,Phenylcarboxylic acid,Phenylformic acid,Purox B,Retarder BA,Retardex,Salvo liquid,Solvo powder,Tenn-Plas,VevoVitall,Tiaprofenic Acid Imp. D (EP) IUPAC Name: benzoic acid SMILES: OC(=O)c1ccccc1
Benzoic Acid, Sigma-Aldrich™
CAS: 65-85-0 Molecular Weight (g/mol): 122.12
Benzoic Acid, SPEX CertiPrep™
HYDRANAL™ - Benzoic acid, Buffer substance for KF titration, Solstice
Buffer substance for Karl Fischer titration
- 1
- 2