Learn More
Benzamide, Sigma-Aldrich™
Product Code. 15864061 Shop All Merck Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864061 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15864061 Supplier Sigma Aldrich Supplier No. 8021910250
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55-21-0
Grade Research
Molecular Weight (g/mol) 121.14
Specifications
Melting Point 124°C to 127°C
Density 1.34 g/cm3 at 20°C
MDL Number MFCD00007968
Quantity 250 g
Synonym Benzamide, Benzoic acid amide, Benzamide