accessibility menu, dialog, popup

UserName

H2NO

CAS RN 55-21-0

IUPAC Name: benzamide
Synonyms: benzamide amid kyseliny benzoove benzamide (acd/name 4.0) 苯唑拉胺 benzoylamide PhC(=O)NH2 PhC(O)NH2
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: KXDAEFPNCMNJSK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)