Learn More
MedChemExpress Bakuchiol
Product Code. 16468908
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16468908 Supplier MedChemExpress Supplier No. HYN0235
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
MedChemExpress Bakuchiol is a phytoestrogen isolated from the seeds of Psoralea corylifolia L; has anti-tumor effects.
Specifications
Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 256.38 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 10309-37-2
Product Type Natural Products
Purity 95.0%
Solubility DMSO : 62.5 mg/mL (243.78 mM∣Need ultrasonic)
Synonym (S)-(+)-Bakuchiol
Target p38 MAPK∣Autophagy