missing translation for 'onlineSavingsMsg'
Learn More

Baeocystin, TRC

Product Code. 30133785
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133785 50 mg 50mg
30004856 5 mg 5mg
30011057 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133785 Supplier Toronto Research Chemicals Supplier No. TRCB11531550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21420-58-6
Molecular Formula C11 H15 N2 O4 P
Molecular Weight (g/mol) 270.222
Synonym N-desmethyl Psilocybin, 3-(2-(methylamino)ethyl)-1H-indol-4-yl, dihydrogen phosphate, 1H-Indol-4-ol, 3-[2-(methylamino)ethyl]-, dihydrogen phosphate (ester) (9CI), Indol-4-ol, 3-[2-(methylamino)ethyl]-, dihydrogen phosphate (ester) (8CI), Baeocystin, Beocystin, [3-[2-(Methylamino)ethyl]-1H-indol-4-yl] dihydrogen phosphate, 1H-Indol-4-ol, 3-[2-(methylamino)ethyl]-, 4-(dihydrogen phosphate), 3-[2-(methylamino)ethyl]-1H-indol-4-ol 4-(dihydrogen phosphate)
IUPAC Name [3-[2-(methylamino)ethyl]-1H-indol-4-yl] dihydrogen phosphate
SMILES CNCCc1c[nH]c2cccc(OP(=O)(O)O)c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H15N2O4P/c1-12-6-5-8-7-13-9-3-2-4-10(11(8)9)17-18(14,15)16/h2-4,7,12-13H,5-6H2,1H3,(H2,14,15,16)
Formula Weight 270.0769
Percent Purity >95%
Chemical Name or Material Baeocystin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.