CAS RN 21420-58-6
CAS RN 21420-58-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Baeocystin, TRC
CAS: 21420-58-6 Molecular Formula: C11 H15 N2 O4 P Molecular Weight (g/mol): 270.222 Synonym: N-desmethyl Psilocybin,3-(2-(methylamino)ethyl)-1H-indol-4-yl, dihydrogen phosphate,1H-Indol-4-ol, 3-[2-(methylamino)ethyl]-, dihydrogen phosphate (ester) (9CI),Indol-4-ol, 3-[2-(methylamino)ethyl]-, dihydrogen phosphate (ester) (8CI),Baeocystin,Beocystin,[3-[2-(Methylamino)ethyl]-1H-indol-4-yl] dihydrogen phosphate,1H-Indol-4-ol, 3-[2-(methylamino)ethyl]-, 4-(dihydrogen phosphate),3-[2-(methylamino)ethyl]-1H-indol-4-ol 4-(dihydrogen phosphate) IUPAC Name: [3-[2-(methylamino)ethyl]-1H-indol-4-yl] dihydrogen phosphate SMILES: CNCCc1c[nH]c2cccc(OP(=O)(O)O)c12