missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Monoclonal Antibody (JAM1EW), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
175.00 EUR - 447.00 EUR
Specifications
| Antigen | B7-H6 |
|---|---|
| Clone | JAM1EW |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16390864
|
Affymetrix eBioscience
46-6526-41 |
25 Tests | |||||||
16300874
![]() |
Invitrogen™
46-6526-42 |
100 Tests |
447.00 EUR
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.Specifications
| B7-H6 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q68D85 | |
| NCR3LG1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| JAM1EW | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Mouse | |
| Human | |
| 374383 | |
| B7-H6 recombinant protein | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| NCR3LG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title