missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Monoclonal Antibody (JAM1EW), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-6526-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry
Specifications
| B7-H6 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Q68D85 | |
| NCR3LG1 | |
| B7-H6 recombinant protein | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| JAM1EW | |
| PerCP-eFluor 710 | |
| NCR3LG1 | |
| DKFZp686I21167 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 374383 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction