Learn More
Brady™ B-428 Metallized Polyester Thermal Transfer Labels
Printable, metallized polyester labels for Thermal Transfer Printers.
191.00 EUR - 905.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | UL, CSA and AGA Approved |
Adhesive Type | Permanent Acrylic |
Color | Gray |
Finish | Matte |
Product Code | Brand | Height (Metric) | Labels per Row | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Labels per Row | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
11503714
|
Brady™
030122 |
6.4 mm | 4 | 8.9 mm | 19.1 mm |
423.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11553784
|
Brady™
030217 |
9.5 mm | 3 | 12.1 mm | 25.4 mm |
448.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11533714
|
Brady™
030125 |
11.1 mm | 5 | 13.6 mm | 12.7 mm |
269.00 EUR
1 roll |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||
11563804
|
Brady™
030241 |
12.7 mm | 1 | 15.2 mm | 50.8 mm |
191.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11513784
|
Brady™
030213 |
12.7 mm | 2 | 15.2 mm | 38.1 mm |
900.00 EUR
1 roll |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||
11513724
|
Brady™
030133 |
25.4 mm | 1 | 28.6 mm | 50.8 mm |
335.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11543724
|
Brady™
030136 |
25.4 mm | 1 | 28.6 mm | 76.2 mm |
500.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11583694
|
Brady™
030110 |
31.8 mm | 1 | 34.9 mm | 69.9 mm |
570.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11573724
|
Brady™
030139 |
50.8 mm | 1 | 54 mm | 76.2 mm |
333.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11513814
|
Brady™
030247 |
50.8 mm | 1 | 54 mm | 101.6 mm |
362.00 EUR
1 roll |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
11573694
|
Brady™
030109 |
19.1 mm | 2 | 22.2 mm | 38.1 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Matte finish
- For smooth surfaces
- Heat resistant with permanent adhesive
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Asset Tracking, Circuit Board Labeling, Equipment Labeling, Data and Telecommunications Labeling, General Identification, Inventory Labeling | |
Rectangle | |
0.112 mm |
UL, CSA and AGA Approved | |
Gray | |
Metallized Polyester | |
Non-sterile | |
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers | |
-70°C to +120°C |