missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Azimilide dihydrochloride

Product Code. 18766826
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18766826 10 mg 10mg
18707816 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18766826 Supplier R&D Systems™ Supplier No. 6318/10
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Kv11.1 (hERG) channel blocker

Azimilide dihydrochloride is a Kv11.1 (hERG) channel blocker, blocks rapidly activating and slowly activating components of delayed rectifier K+ currents (IC50 of 0.4 mM and 3 mM, respectively). Also inhibits Na+/Ca2+ exchanger in vitro. Shows inhibition of Na+ currents, L-type Ca2+ currents and other K+ currents at high concentrations.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[[[5-(4-Chlorophenyl)-2-furanyl]methylene]amino]-3-[4-(4-methyl-1-piperazinyl)butyl]-2,4-imidazolidinedione dihydrochloride
CAS 149888-94-8
Assay Percent Range >97%
Quantity 10 mg
Product Line Tocris
Molecular Formula C23H28ClN5O3.2HCl
Formula Weight Observed MW: 530.88
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.