missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZD 5438

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent inhibitor of cyclin dependent kinases (cdks; reported IC50 values are 6 - 45, 14, 16 and 20 nM for cdk2, cdk5, cdk1 and cdk9 respectively). Also inhibits cdk4 and cdk7 in the sub micromolar range. Exhibits antiproliferative activity in human tumor cell lines. Blocks cell cycling at G2-M, S and G1 phases; reduces the proportion of actively cycling cells in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 371.46 |
| Purity | >99% |
| Molecular Weight (g/mol) | 371.459 |
| Chemical Name or Material | AZD 5438 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction