CAS RN 602306-29-6
CAS RN 602306-29-6
IUPAC Name:
N-(4-methanesulfonylphenyl)-4-[2-methyl-1-(propan-2-yl)-1H-imidazol-5-yl]pyrimidin-2-amine
Synonyms:
4-(3-isopropyl-2-methylimidazol-4-yl)-N-(4-methanesulfonylphenyl)pyrimidin-2-amine
Molecular Weight (g/mol):
371.46
Molecular Formula:
C18H21N5O2S
InChi Key:
WJRRGYBTGDJBFX-UHFFFAOYSA-N
SMILES:
CC(C)N1C(C)=NC=C1C1=NC(NC2=CC=C(C=C2)S(C)(=O)=O)=NC=C1
1
–
2
of
2
results
1
Tocris Bioscience™ AZD 5438
CAS: 602306-29-6 Molecular Formula: C18H21N5O2S
2
AZD-5438, MedChemExpress
MedChemExpress AZD-5438 is a potent CDK1, CDK2, and CDK9 inhibitor, with IC50s of 16 nM, 6 nM, and 20 nM in cell-free assays, respectively. AZD-5438 shows less inhibition activity against GSK3β, CDK5 and CDK6 .