missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 5438
GREENER_CHOICE

Product Code. 18005242
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18005242

Brand: Tocris Bioscience™ 3968/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk1, 2, 5 and 9

Potent inhibitor of cyclin dependent kinases (cdks; reported IC50 values are 6 - 45, 14, 16 and 20 nM for cdk2, cdk5, cdk1 and cdk9 respectively). Also inhibits cdk4 and cdk7 in the sub micromolar range. Exhibits antiproliferative activity in human tumor cell lines. Blocks cell cycling at G2-M, S and G1 phases; reduces the proportion of actively cycling cells in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 602306-29-6
Molecular Formula C18H21N5O2S

Specifications

Quantity 10 mg
Formula Weight 371.46
Purity >99%
Molecular Weight (g/mol) 371.459
Chemical Name or Material AZD 5438
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.